1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-Heptadecafluoro-10-iododecane ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 1H,1H,2H,2H-Perfluoro-1-iododecane, 1H,1H,2H,2H-Perfluorodecyliodide, 1-Iodo-1H,1H,2H,2H-perfluorodecane

09-5972-25G 09-5972-5G
100209-616EA 474.15 USD
100209-616 100209-618
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-Heptadecafluoro-10-iododecane ≥97%
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-Heptadecafluoro-10-iododecane

Organo Fluorine

Formula: C₁₀H₄F₁₇I
MW: 574.02 g/mol
Boiling Pt: 178…180 °C
Melting Pt: 55.5…56.5 °C
Density: 1.88 g/cm³ (60 °C)
Storage Temperature: Ambient
MDL Number: MFCD00039411
CAS Number: 2043-53-0
EINECS: 218-053-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR