1H,1H,2H,2H-Heptadecafluoro-1-decanol ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluoro-1-decanol, 1H,1H,2H,2H-Perfluoro-1-decanol, 1H,1H,2H,2H-Perfluorodecan-1-ol

09-5969-100G 09-5969-25G
100203-936EA 533.6 USD
100203-936 100207-546
1H,1H,2H,2H-Heptadecafluoro-1-decanol ≥97%
1H,1H,2H,2H-Heptadecafluoro-1-decanol

Organo Fluorine

Formula: C₁₀H₅F₁₇O
MW: 464.12 g/mol
Melting Pt: 40…43 °C
MDL Number: MFCD00039544
CAS Number: 678-39-7
EINECS: 211-648-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR