Pentafluoropyridine ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 2,3,4,5,6-Perfluoropyridine, 2,3,4,5,6-Pentafluoropyridine

09-5800-25G 09-5800-5G
100203-716EA 359.51 USD
100203-716 100203-932
Pentafluoropyridine ≥99%
Pentafluoropyridine

Organo Fluorine

Formula: C₅F₅N
MW: 169.05 g/mol
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: -41 °C
Density: 1.54 g/cm³ (20 °C)
Flash Pt: 75 °F
Storage Temperature: Ambient
MDL Number: MFCD00006225
CAS Number: 700-16-3
EINECS: 211-839-9
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR