Pentafluorophenol ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 2,3,4,5,6-Pentafluorophenol

09-5630-25G 09-5630-5G
100197-992EA 242.03 USD
100197-992 100196-008
Pentafluorophenol ≥99%
Pentafluorophenol

Organo Fluorine

Formula: C₆HF₅O
MW: 184.07 g/mol
Boiling Pt: 143 °C
Melting Pt: 26…29 °C
Density: 1.75 g/cm³ (20 °C)
Flash Pt: 162 °F
Storage Temperature: Ambient
MDL Number: MFCD00002156
CAS Number: 771-61-9
EINECS: 212-235-8
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR