Pentafluorobenzyl bromide ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
α-Bromo-2,3,4,5,6-pentafluorotoluene, 2,3,4,5,6-Pentafluorobenzyl bromide
09-5550-25G
09-5550-1G
09-5550-5G
100199-744EA
447.26
USD
100199-744
100206-270
100206-272
Pentafluorobenzyl bromide ≥98%
Pentafluorobenzyl bromide
Organo Fluorine
Formula:
(C₆F₅)CH₂Br MW: 260.99 g/mol Boiling Pt: 174 Density: 1.728 Flash Pt: 181 °F Storage Temperature: Ambient |
MDL Number:
MFCD00000299 CAS Number: 1765-40-8 EINECS: 217-182-4 UN: 1760 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...