2,3,4,5,6-Pentafluoroaniline ≥99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
09-5420-25G
09-5420-5G
100204-866EA
322.71
USD
100204-866
100204-868
2,3,4,5,6-Pentafluoroaniline ≥99%
2,3,4,5,6-Pentafluoroaniline
Organo Fluorine
Formula:
C₆H₂F₅N Boiling Pt: 153 °C Melting Pt: 34 °C Density: 1,744 g/cm³ (20 °C) Flash Pt: 165 °F Storage Temperature: Ambient |
MDL Number:
MFCD00007643 CAS Number: 771-60-8 EINECS: 212-234-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...