Pentadecafluorocaprylic acid ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Pentadecafluorooctoic acid, Pentadecafluorooctanoic acid, PFOA, Perfluorocaprylic acid, Perfluorooctanoic acid
09-5380-5G
09-5380-25G
100199-742EA
72.18
USD
100199-742
100208-778
Pentadecafluorocaprylic acid ≥98%
Pentadecafluorocaprylic acid
Organo Fluorine
Formula:
C₈HF₁₅O₂ MW: 414.07 g/mol Boiling Pt: 189 °C (736 mmHg) Melting Pt: 59…60 °C |
MDL Number:
MFCD00004174 CAS Number: 335-67-1 EINECS: 206-397-9 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...