1H,1H,9H-Hexadecafluoro-1-nonanol ≥95%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-Hexadecafluoro-1-nonanol, 1H,1H,9H-Perfluoro-1-nonanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-Hexadecafluor-1-nonanol, 1H,1h,9h-hexadecafluorononanol

09-4530-10G 09-4530-50G
100198-094EA 205.23 USD
100198-094 100204-492
1H,1H,9H-Hexadecafluoro-1-nonanol ≥95%
1H,1H,9H-Hexadecafluoro-1-nonanol

Organo Fluorine

Formula: C₉H₄F₁₆O
MW: 432.1 g/mol
MDL Number: MFCD00039629
CAS Number: 376-18-1
EINECS: 206-806-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR