4-Fluorophenol ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: p-Fluorophenol, 1-Fluoro-4-hydroxybenzene

09-4090-100G 09-4090-25G
100207-338EA 253.35 USD
100207-338 100202-900
4-Fluorophenol ≥99%
4-Fluorophenol

Organo Fluorine

Formula: C₆H₅FO
MW: 112.1 g/mol
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 44 - 47 °C
Density: 1.22 g/cm³ (20 °C)
Flash Pt: 68 °C
Storage Temperature: Ambient
MDL Number: MFCD00002316
CAS Number: 371-41-5
EINECS: 206-736-0
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR