2-Fluorophenol ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
09-4070-10G
09-4070-50G
100199-740EA
73.6
USD
100199-740
100207-960
2-Fluorophenol ≥98%
2-Fluorophenol
Organo Fluorine
Formula:
C₆H₅FO Boiling Pt: 171…172 °C (741 mmHg) Melting Pt: 16.1 °C Density: 1.256 Flash Pt: 116 °F Storage Temperature: Ambient |
MDL Number:
MFCD00002155 CAS Number: 367-12-4 EINECS: 206-681-2 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...