2-Fluorobenzoic acid 97
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
o-Fluorobenzoic acid
09-3500-25G
09-3500-100G
100196-004EA
67.94
USD
100196-004
100204-848
2-Fluorobenzoic acid 97
2-Fluorobenzoic acid
Organo Fluorine
Formula:
C₇H₅FO₂ MW: 140.11 g/mol Boiling Pt: 245 Melting Pt: 123…125 °C Density: 1.46 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002405 CAS Number: 445-29-4 EINECS: 207-158-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...