3-Chloro-4-fluorobenzotrifluoride ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 1-Chloro-2-fluoro-5-(trifluoromethyl)benzene, 3-Chloro-4-fluorobenzotrifluoride, 2-Chloro-1-fluoro-4-(trifluoromethyl)benzene

09-1863-10G 09-1863-50G
100198-552EA 369.42 USD
100198-552 100204-486
3-Chloro-4-fluorobenzotrifluoride ≥98%
3-Chloro-4-fluorobenzotrifluoride

Organo Fluorine

Formula: C₇H₃ClF₄
MW: 198.55 g/mol
Density: 1,490
MDL Number: MFCD00042207
CAS Number: 78068-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR