3-Bromo-1,1,1-trifluoroacetone ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

09-1600-25G 09-1600-5G
100203-712EA 485.48 USD
100203-712 100209-580
3-Bromo-1,1,1-trifluoroacetone ≥97%
3-Bromo-1,1,1-trifluoroacetone

Organo Fluorine

Formula: BrH₂CC(O)CF₃
MW: 190.95 g/mol
Boiling Pt: 87 °C (1013 hPa)
Density: 1.8389 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00039237
CAS Number: 431-35-6
EINECS: 207-071-9
UN: 2924
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR