Bromopentafluorobenzene ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 1-Bromo-2,3,4,5,6-pentafluorobenzene

09-1520-50G 09-1520-10G
100197-054EA 332.62 USD
100197-054 100200-396
Bromopentafluorobenzene ≥99%
Bromopentafluorobenzene

Organo Fluorine

Formula: C₆BrF₅
MW: 246.96 g/mol
Boiling Pt: 137 °C (1013 hPa)
Melting Pt: -31 °C
Density: 1.9503 g/cm³ (20 °C)
Flash Pt: 190 °F
Storage Temperature: Ambient
MDL Number: MFCD00000287
CAS Number: 344-04-7
EINECS: 206-449-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR