4,4,4-Trifluoro-1-phenyl-1,3-butanedione ≥98%
Supplier: STREM CHEMICALS INC MS
Synonyms:
3-Benzoyl-1,1,1-trifluoroacetone, (4,4,4-Trifluor-1,3-butandionyl)benzol, 4,4,4-Trifluoro-1-phenylbutane-1,3-dione, Benzoyl-1,1,1-trifluoroacetone
09-1210-50G
09-1210-10G
100198-086EA
186.83
USD
100198-086
100195-998
4,4,4-Trifluoro-1-phenyl-1,3-butanedione ≥98%
4,4,4-Trifluoro-1-phenyl-1,3-butanedione
Organo Fluorine
Formula:
C₁₀H₇F₃O₂ MW: 216.16 g/mol Boiling Pt: 224 °C Melting Pt: 38…40 °C |
MDL Number:
MFCD00000425 CAS Number: 326-06-7 EINECS: 206-307-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...