4-Chloro-3-(trifluoromethyl)aniline ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 5-Amino-2-chloro-α,α,α-trifluorotoluene, 1-Amino-4-chloro-3-(trifluoromethyl)benzene, 4-Chloro-α,α,α-trifluoro-m-toluidine, 5-Amino-2-chlorobenzotrifluoride

09-1070-5G 09-1070-25G
100204-822EA 43.88 USD
100204-822 100207-952
4-Chloro-3-(trifluoromethyl)aniline ≥97%
4-Chloro-3-(trifluoromethyl)aniline

Organo Fluorine

Formula: C₇H₅ClF₃N
MW: 195.57 g/mol
Boiling Pt: 130…132 °C (27 mmHg)
Melting Pt: 36…37 °C
Flash Pt: >230 °F
Storage Temperature: Ambient
MDL Number: MFCD00007846
CAS Number: 320-51-4
EINECS: 206-277-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR