(S,S)-(+)-2,5-Hexanediol ≥99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
(2S,5S)-2,5-Hexanediol, (2S,5S)-Hexane-2,5-diol, (2S,5S)-(+)-2,5-Hexanediol, (S,S)-(+)-2,5-Dihydroxy hexane
08-2025-500MG
08-2025-2G
100200-838EA
130.22
USD
100200-838
100202-618
(S,S)-(+)-2,5-Hexanediol ≥99%
(S,S)-(+)-2,5-Hexanediol
Formula:
C₆H₁₄O₂ MW: 118.18 g/mol Melting Pt: 50…53 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00082583 CAS Number: 34338-96-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...