(R,R)-(-)-2,5-Hexanediol ≥99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
08-2024-2G
08-2024-500MG
100202-616EA
393.48
USD
100202-616
100200-836
(R,R)-(-)-2,5-Hexanediol ≥99%
(R,R)-(-)-2,5-Hexanediol
Formula:
C₆H₁₄O₂ Melting Pt: 50…53 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00142339 CAS Number: 17299-07-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...