5,5-Dimethylacetylacetone ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 5,5-Dimethylhexane-2,4-dione, 2,2-Dimethyl-3,5-hexanedione, 5,5-Diméthylhexanedione-2,4

08-2000-25G 08-2000-5G
100208-286EA 1068.62 USD
100208-286 100208-288
5,5-Dimethylacetylacetone ≥97%
5,5-Dimethylacetylacetone
Formula: C₈H₁₄O₂
MW: 142.2 g/mol
MDL Number: MFCD00142767
CAS Number: 7307-04-2
EINECS: 230-760-0
UN: 1224
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR