1,1,1,5,5,5-Hexafluoroacetylacetone ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Hexafluoroacetylacetone, 1,1,1,5,5,5-Hexafluoropentane-2,4-dione, 1,1,1,5,5,5-Hexafluoro-2,4-pentanedione, HFAA

08-0750-5G 08-0750-100G 08-0750-25G
100197-336EA 52.37 USD
100197-336 100200-630 100200-632
1,1,1,5,5,5-Hexafluoroacetylacetone ≥98%
1,1,1,5,5,5-Hexafluoroacetylacetone
Formula: C₅H₂F₆O₂
MW: 208.06 g/mol
Boiling Pt: 70 °C
Density: 1.47 g/cm³ (20 °C)
Flash Pt: 32 °C
Storage Temperature: Ambient
MDL Number: MFCD00000426
CAS Number: 1522-22-1
EINECS: 216-191-0
UN: 2920
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR