18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane) ≥99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
1,4,7,10,13,16-Hexaoxacyclooctadecane, 18-Crown-6
08-0320-50G
08-0320-10G
100210-782EA
393.48
USD
100210-782
100203-908
18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane) ≥99%
18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane)
Formula:
C₁₂H₂₄O₆ MW: 264.32 g/mol Boiling Pt: 396 °C (1013 hPa) Melting Pt: 37 - 39 °C Density: 1.237 g/cm³ (25 °C) Flash Pt: 109 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005113 CAS Number: 17455-13-9 EINECS: 241-473-5 Merck Index: 13,02630 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...