(S)-(+)-1,3-Butanediol ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (S)-(+)-1,3-Butylene glykol, (S)-Butane-1,3-diol, (S)-(+)-1,3-Dihydroxy butane

08-0198-250MG 08-0198-1G
100198-082EA 104.74 USD
100198-082 100207-528
(S)-(+)-1,3-Butanediol ≥98%
(S)-(+)-1,3-Butanediol
Formula: C₄H₁₀O₂
MW: 90.12 g/mol
Boiling Pt: 109 °C (1013 hPa)
Density: 1 g/cm³ (23 °C)
Flash Pt: 249 °F
MDL Number: MFCD00064278
CAS Number: 24621-61-2
EINECS: 246-363-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR