Osmium ≥99.8%, powder
Supplier: STREM CHEMICALS INC MS
|
Danger
|
76-3000-5G
76-3000-1G
100209-268EA
1002.09
USD
100209-268
100199-984
Osmium ≥99.8%, powder
Osmium
Formula:
Os Boiling Pt: 5027 °C Melting Pt: 3000 °C Density: 22,48 |
MDL Number:
MFCD00011147 CAS Number: 7440-04-2 EINECS: 231-114-0 UN: 3089 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...