Osmium tetraoxide 4% in water
Supplier: STREM CHEMICALS INC MS
|
Danger
|
76-2952-5ML
76-2952-2ML
76-2952-10ML
100196-360EA
106.15
USD
100196-360
100197-596
100201-246
Osmium tetraoxide 4% in water
Osmium tetraoxide
Formula:
O₄Os Storage Temperature: Refrigerator |
MDL Number:
MFCD00011150 CAS Number: 20816-12-0 EINECS: 244-058-7 UN: 2471 ADR: 6.1,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...