Diammonium hexabromoosmate ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

76-0080-5G 76-0080-1G
100197-594EA 938.4 USD
100197-594 100197-592
Diammonium hexabromoosmate ≥99%
Diammonium hexabromoosmate
Formula: H₈Br₆N₂Os
MDL Number: MFCD00049389
CAS Number: 24598-62-7
EINECS: 246-340-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR