Rhenium trioxide ≥99.9% (Re basis)

Supplier: STREM CHEMICALS INC MS

Synonyms: Rhenium(VI) oxide, Rhenium (VI) oxide

75-2500-5G 75-2500-1G
100198-880EA 644 USD
100198-880 100198-878
Rhenium trioxide ≥99.9% (Re basis)
Rhenium trioxide
Formula: O₃Re
MW: 234.21 g/mol
Melting Pt: 400 °C (dec.)
Density: 7.4 g/cm³ (20 °C)
MDL Number: MFCD00016306
CAS Number: 1314-28-9
EINECS: 215-228-8
Merck Index: 12,08347

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR