Rhenium ≥99.99%, powder, PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

75-1890-5G 75-1890-1G
100201-730EA 352.43 USD
100201-730 100201-728
Rhenium ≥99.99%, powder, PURATREM
Rhenium
Formula: Re
MW: 186.21 g/mol
Boiling Pt: 5630 °C (1013 hPa)
Melting Pt: 3180 °C
Density: 21.02 g/cm³ (20 °C)
MDL Number: MFCD00011195
CAS Number: 7440-15-5
EINECS: 231-124-5
UN: 3089
ADR: 4.1,II
Merck Index: 12,08343

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR