Tungsten(VI) chloride ≥99.9% (W basis)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Tungsten hexachloride
74-2350-500G
74-2350-25G
74-2350-100G
100199-980EA
1321.97
USD
100199-980
100211-160
100199-978
Tungsten(VI) chloride ≥99.9% (W basis)
Tungsten(VI) chloride
Formula:
Cl₆W MW: 396.56 g/mol Boiling Pt: 346.7 °C Melting Pt: 275 °C Density: 3.518 g/cm³ (20 °C) |
MDL Number:
MFCD00011463 CAS Number: 13283-01-7 EINECS: 236-293-9 UN: 1759 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...