Tungsten ≥99.98%, rod annealed

Supplier: STREM CHEMICALS INC MS
Warning

74-0075-500CM 74-0075-100CM 74-0075-25CM
100200-296EA 2605.72 USD
100200-296 100201-724 100210-056
Tungsten ≥99.98%, rod annealed
Tungsten
Formula: W
MW: 183.84 g/mol
Boiling Pt: 5660
Melting Pt: 3410
Density: 19.3
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9
Merck Index: 12,09945

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR