Tantalum(V) oxide ≥99.8% (99.95+%-Ta)

Supplier: STREM CHEMICALS INC MS

73-2000-25G 73-2000-100G
100196-346EA 128.8 USD
100196-346 100196-344
Tantalum(V) oxide ≥99.8% (99.95+%-Ta)
Tantalum(V) oxide
Formula: O₅Ta₂
Melting Pt: 1800 °C
Density: 8.2
MDL Number: MFCD00011254
CAS Number: 1314-61-0
EINECS: 215-238-2
Merck Index: 14,09056

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR