Tantalum pentachloride, anhydrous ≥99.9% (Ta basis)

Supplier: STREM CHEMICALS INC MS
Danger

73-1000-10G 73-1000-50G
100198-872EA 76.43 USD
100198-872 100198-874
Tantalum pentachloride, anhydrous ≥99.9% (Ta basis)
Tantalum pentachloride
Formula: Cl₅Ta
Boiling Pt: 232 - 235 °C (1013 hPa)
Melting Pt: 216 - 217 °C
Density: 3.68 g/cm³ (20 °C)
MDL Number: MFCD00011253
CAS Number: 7721-01-9
EINECS: 231-755-6
UN: 1759
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR