Tantalum carbide ≥99.5% (Ta basis)
Supplier: STREM CHEMICALS INC MS
Synonyms:
Tantalum(IV) carbide
73-0750-100G
73-0750-25G
73-0750-5G
100201-244EA
519.45
USD
100201-244
100202-508
100204-100
Tantalum carbide ≥99.5% (Ta basis)
Tantalum carbide
Formula:
CTa MW: 192.96 Boiling Pt: 5500 Melting Pt: 3880 |
MDL Number:
MFCD00011255 CAS Number: 12070-06-3 EINECS: 235-118-3 UN: 3178 ADR: 4.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...