Tantalum ≥99.95%, rods
Supplier: STREM CHEMICALS INC MS
|
Warning
|
73-0065-5CM
73-0065-25CM
73-0065-1CM
100202-686EA
778.46
USD
100202-686
100197-588
100198-242
Tantalum ≥99.95%, rods
Tantalum
Formula:
Ta Boiling Pt: 5425 °C Melting Pt: 2996 °C Density: 16.6 |
MDL Number:
MFCD00011252 CAS Number: 7440-25-7 EINECS: 231-135-5 Merck Index: 12,09223 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...