Tantalum ≥99.95%, foil

Supplier: STREM CHEMICALS INC MS
Warning

73-0050-200X200MM 73-0050-100X100MM 73-0050-50X50MM
100206-128EA 2474.09 USD
100206-128 100203-806 100203-268
Tantalum ≥99.95%, foil
Tantalum
Formula: Ta
Boiling Pt: 5425 °C
Melting Pt: 2996 °C
Density: 16.69 g/cm³ (20 °C)
MDL Number: MFCD00011252
CAS Number: 7440-25-7
EINECS: 231-135-5
Merck Index: 12,09223

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR