Hafnium ≥99.5% (Zr-2.9%), foil

Supplier: STREM CHEMICALS INC MS

72-0075-100X100MM 72-0075-50X50MM
100200-294EA 1207.32 USD
100200-294 100199-256
Hafnium ≥99.5% (Zr-2.9%), foil
Hafnium
Formula: Hf
MW: 178.49 g/mol
Boiling Pt: 4602 °C
Melting Pt: 2227 °C
Density: 13,31
MDL Number: MFCD00011032
CAS Number: 7440-58-6
EINECS: 231-166-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR