Lutetium ≥99.9%, ingot

Supplier: STREM CHEMICALS INC MS
Warning

71-0100-1G 71-0100-5G
100208-946EA 351.02 USD
100208-946 100208-948
Lutetium ≥99.9%, ingot
Lutetium
Formula: Lu
MW: 174.97 g/mol
Boiling Pt: 3000 °C (1013 hPa)
Melting Pt: 1651 - 1675 °C
Density: 9.849 g/cm³ (25 °C)
MDL Number: MFCD00011098
CAS Number: 7439-94-3
EINECS: 231-103-0
Merck Index: 12,05642

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR