EDTA dipotassium salt dihydrate ≥99%
Supplier: STREM CHEMICALS INC MS
Synonyms:
Ethylenediaminetetraacetic acid dipotassium salt dihydrate, Titriplex® Potassium salt, Ethylenediaminetetra-acetic acid dipotassium salt dihydrate, Edetic acid dipotassium salt dihydrate, Dipotassium dihydrogen ethylenediaminetetraacetate dihydrate, Ethylenedinitrilotetra-acetic acid dipotassium salt dihydrate, EDTA dipotassium dihydrate, Potassium 2,2'-((2-(bis(carboxymethyl)amino)ethyl)azanediyl)diacetate dihydrate, Ethylenedinitrilotetraacetic acid dipotassium salt
07-0580-50G
07-0580-250G
100208-270EA
65.11
USD
100208-270
100203-706
EDTA dipotassium salt dihydrate ≥99%
EDTA dipotassium salt dihydrate
Formula:
C₁₀H₁₄K₂N₂O₈·2H₂O MW: 404.46 g/mol Melting Pt: 272 (dec.) Density: 1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00150036 CAS Number: 25102-12-9 EINECS: 217-895-0 Merck Index: 13,03546 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...