(1S,2S)-(+)-1,2-Diaminocyclohexane ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

07-0335-1G 07-0335-5G
100202-482EA 101.91 USD
100202-482 100202-354
(1S,2S)-(+)-1,2-Diaminocyclohexane ≥99%
(1S,2S)-(+)-1,2-Diaminocyclohexane
Formula: C₆H₁₄N₂
Boiling Pt: 65 °C (5 mmHg)
Melting Pt: 38…40 °C
Flash Pt: 169 °F
Storage Temperature: Refrigerator
MDL Number: MFCD00062986
CAS Number: 21436-03-3
UN: 3259
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR