(1R,2R)-(-)-1,2-Diaminocyclohexane ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: (R,R)-DACH, (1R,2R)-Cyclohexane-1,2-diamine

07-0330-5G 07-0330-1G
100199-722EA 356.68 USD
100199-722 100206-230
(1R,2R)-(-)-1,2-Diaminocyclohexane ≥99%
(1R,2R)-(-)-1,2-Diaminocyclohexane
Formula: C₆H₁₄N₂
MW: 114.19 g/mol
Boiling Pt: 65 °C (5 mmHg)
Melting Pt: 38…40 °C
Flash Pt: 169 °F
Storage Temperature: Refrigerator
MDL Number: MFCD00062985
CAS Number: 20439-47-8
UN: 3259
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR