trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Titriplex® IV
07-0318-25G
07-0318-100G
100206-226EA
77.85
USD
100206-226
100197-990
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate ≥98%
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate
Formula:
C₁₄H₂₂N₂O₈·1H₂O MW: 364.35 g/mol Melting Pt: > 210 °C |
MDL Number:
MFCD00066429 CAS Number: 125572-95-4 EINECS: 236-308-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...