Erbium trichloride hydrate ≥99.999%, PURATREM (low Ca, Fe, Mg)

Supplier: STREM CHEMICALS INC MS
Warning

68-6805-50G 68-6805-10G
100199-428EA 932.74 USD
100199-428 100210-038
Erbium trichloride hydrate ≥99.999%, PURATREM (low Ca, Fe, Mg)
Erbium trichloride hydrate
Formula: Cl₃Er·H₂O
MW: 291.63 g/mol
Melting Pt: 168 °C
MDL Number: MFCD00010989
CAS Number: 19423-85-9
EINECS: 233-385-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR