Erbium trichloride hydrate ≥99.999%, PURATREM

Supplier: STREM CHEMICALS INC MS
Warning

68-6804-10G 68-6804-50G
100208-944EA 175.51 USD
100208-944 100204-638
Erbium trichloride hydrate ≥99.999%, PURATREM
Erbium trichloride hydrate
Formula: Cl₃Er·H₂O
MW: 291.63 g/mol
Melting Pt: 168 °C
MDL Number: MFCD00010989
CAS Number: 19423-85-9
EINECS: 233-385-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR