Tetramethyl(propyl)cyclopentadiene ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Tetramethyl(n-propyl)cyclopentadiene

06-3050-25G 06-3050-1G 06-3050-5G
100197-310EA 736 USD
100197-310 100201-334 100197-312
Tetramethyl(propyl)cyclopentadiene ≥97%
Tetramethyl(propyl)cyclopentadiene

Metallocenes, Derivatives & Cp Precursors

Formula: C₁₂H₂₀
MW: 164.29 g/mol
MDL Number: MFCD00145386
CAS Number: 64417-12-5
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR