1,2,3,4,5-Pentaphenylcyclopentadiene ≥99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
1,2,3,4,5-Pentaphenyl-1,3-cyclopentadiene, Cyclopenta-1,3-diene-1,2,3,4,5-pentaylpentabenzene, Pentaphenylcyclopentadiene
06-1296-1G
06-1296-5G
100206-214EA
124.55
USD
100206-214
100206-216
1,2,3,4,5-Pentaphenylcyclopentadiene ≥99%
1,2,3,4,5-Pentaphenylcyclopentadiene
Metallocenes, Derivatives & Cp Precursors
Formula:
C₃₅H₂₆ MW: 446.59 g/mol Melting Pt: 254…256 °C |
MDL Number:
MFCD00012263 CAS Number: 2519-10-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...