1,2,3,4,5-Pentamethylcyclopentadiene ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Pentamethylcyclopentadiene, 1,2,3,4,5-Pentamethylcyclopenta-1,3-diene, 1,2,3,4,5-Pentamethyl-1,3-cyclopentadiene

06-1290-25G 06-1290-5G 06-1290-1G
100195-954EA 635.51 USD
100195-954 100209-512 100209-510
1,2,3,4,5-Pentamethylcyclopentadiene ≥98%
1,2,3,4,5-Pentamethylcyclopentadiene

Metallocenes, Derivatives & Cp Precursors

Formula: C₁₀H₁₆
MW: 136.24 g/mol
Boiling Pt: 58.3 °C (13.5 mmHg)
Density: 0.87 g/cm³ (20 °C)
Flash Pt: 112 °F
Storage Temperature: Ambient
MDL Number: MFCD00001354
CAS Number: 4045-44-7
EINECS: 223-743-4
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR