1-Chloro-2,2-dimethylpropane ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Chloro-1-diméthyl-2,2-propane, neo-Pentylchloride

06-1150-100G 06-1150-25G 06-1150-5G
100200-232EA 791.2 USD
100200-232 100203-892 100203-894
1-Chloro-2,2-dimethylpropane ≥98%
1-Chloro-2,2-dimethylpropane
Formula: C₅H₁₁Cl
MW: 106.6 g/mol
Boiling Pt: 84
Melting Pt: -20
Density: 0.866
Flash Pt: 16 °F
MDL Number: MFCD00000941
CAS Number: 753-89-9
EINECS: 212-040-8
UN: 1993
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR