Neodymium(III) chloride hydrate ≥99.997% (Nd basis), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Neodymium trichloride hydrate
60-6030-10G
60-6030-50G
100200-712EA
117.48
USD
100200-712
100199-968
Neodymium(III) chloride hydrate ≥99.997% (Nd basis), PURATREM
Neodymium(III) chloride hydrate
Formula:
NdCl₃·xH₂O MW: 250.60 g/mol (anhy) Melting Pt: 123 °C Density: 2.28 g/cm³ (20 °C) |
MDL Number:
MFCD00011132 CAS Number: 13477-89-9 EINECS: 233-031-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...