Diphenylacetylene ≥99%

Supplier: STREM CHEMICALS INC MS

06-0550-2G 06-0550-10G
100204-780EA 45.29 USD
100204-780 100209-508
Diphenylacetylene ≥99%
Diphenylacetylene
Formula: C₁₄H₁₀
MW: 178.23 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 56 - 59 °C
Density: 0.99 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00004786
CAS Number: 501-65-5
EINECS: 207-926-6
Merck Index: 12,09643

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR