Carbon black ≥99.99%, acetylene, 50% compressed

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Furnace black, Lamp Black

06-0026-1000G 06-0026-250G
100206-206EA 298.65 USD
100206-206 100201-324
Carbon black ≥99.99%, acetylene, 50% compressed
Carbon black
Formula: C
MW: 12.01 g/mol
Boiling Pt: 4200 °C
Melting Pt: 3652…3697 °C (subl.)
Density: 0.10 g/cm³ (bulk density)
MDL Number: MFCD00133992
CAS Number: 1333-86-4
EINECS: 231-153-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR