Cerium triacetate hydrate ≥99.999% (Ce basis), PURATREM
Supplier: STREM CHEMICALS INC MS
Synonyms:
Acetic acid cerium (III) salt hydrate, Cerium (III) acetate hydrate
58-5801-100G
58-5801-25G
100198-866EA
736
USD
100198-866
100210-028
Cerium triacetate hydrate ≥99.999% (Ce basis), PURATREM
Cerium triacetate hydrate
Formula:
C₆H₉CeO₆·H₂O MW: 335.266 g/mol |
MDL Number:
MFCD00150533 CAS Number: 206996-60-3 EINECS: 208-654-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...